Assay Details
| Gene Symbol: TPPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015964.3 | NP_057048.2 | 1 - 2 | 160 | 85 | |
| GenBank mRNA | AF132972.1 | - | 1 - 2 | 80 | 85 | |
| BC000691.2 | - | 1 - 2 | 107 | 3349849 | 85 | |
| BQ640401.1 | - | 1 - 2 | 160 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001187.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF132972.1,BC000691.2,NM_015964.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016525.hg.1 | ENST00000290942.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016525.hg.1 | ENST00000393957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016525.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016525.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016525.hg.1 | TC16001187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016527.hg.1 | ENST00000393957.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016527.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016527.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016527.hg.1 | TC16001187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000458950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181905.hg.1 | TC1600010627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181902.hg.1 | TC1600010627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000290942.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000393957.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000458950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000561537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000562206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | ENST00000564104.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_015964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010627.hg.1 | NM_016140.3 | Refseq |

TPPP3
Genomic Map