Assay Details
| Gene Symbol: MFAP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021647.7 | NP_067679.6 | 2 - 3 | 627 | 99 | |
| XM_005263366.3 | XP_005263423.1 | 3 - 4 | 561 | 99 | ||
| XM_017008864.1 | XP_016864353.1 | 4 - 5 | 505 | 99 | ||
| XM_017008865.1 | XP_016864354.1 | 4 - 5 | 3621 | 99 | ||
| XM_017008866.1 | XP_016864355.1 | 4 - 5 | 695 | 99 | ||
| XM_017008867.1 | XP_016864356.1 | 4 - 5 | 501 | 99 | ||
| XM_017008868.1 | XP_016864357.1 | 3 - 4 | 776 | 99 | ||
| XM_017008869.1 | XP_016864358.1 | 2 - 3 | 3608 | 99 | ||
| XM_017008870.1 | XP_016864359.1 | 2 - 3 | 3568 | 99 | ||
| XM_017008871.1 | XP_016864360.1 | 2 - 3 | 508 | 99 | ||
| GenBank mRNA | AB014526.1 | - | 2 - 3 | 475 | 99 | |
| AK290041.1 | - | 2 - 3 | 537 | 99 | ||
| AY391838.1 | - | 1 - 2 | 408 | 99 | ||
More Information
Set Membership: |
|

MFAP3L
Genomic Map