| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05015125.hg.1 |
TC05001037.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05015125.hg.1 |
ENST00000361362.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05015125.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05015123.hg.1 |
TC05001037.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05015123.hg.1 |
ENST00000361362.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05015123.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520301.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
ENST00000523286.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009671.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500168102.hg.1 |
TC0500009671.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
ENST00000361362.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
ENST00000520660.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
ENST00000520805.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
NM_030613.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500168104.hg.1 |
TC0500009671.hg.1 |
Clariom D TC |