Assay Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030613.3 | NP_085116.2 | 3 - 4 | 307 | 112 | |
| GenBank mRNA | AK131367.1 | - | 3 - 4 | 286 | 112 | |
| BC132781.1 | - | 1 - 2 | 104 | 40146803 | 112 | |
| BC142989.1 | - | 3 - 4 | 275 | 8860436 | 112 | |
| DA694775.1 | - | 3 - 4 | 286 | 112 | ||
| DB059183.1 | - | 3 - 4 | 307 | 112 | ||
| Gene Symbol: LOC107984119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010140.1 | XP_016865629.1 | 3 - 4 | 567 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001037.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK131367.1,BC132781.1,BC142989.1,NM_030613.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015133.hg.1 | TC05001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015133.hg.1 | ENST00000361362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015133.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015133.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015138.hg.1 | TC05001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015138.hg.1 | ENST00000361362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015138.hg.1 | ENST00000503510.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015138.hg.1 | ENST00000520301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015138.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015138.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015135.hg.1 | TC05001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015135.hg.1 | ENST00000361362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015135.hg.1 | ENST00000503510.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015135.hg.1 | ENST00000520301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015135.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015135.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015131.hg.1 | TC05001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015131.hg.1 | ENST00000361362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015131.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015131.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015137.hg.1 | TC05001037.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015137.hg.1 | ENST00000361362.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015137.hg.1 | ENST00000503510.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015137.hg.1 | ENST00000520301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015137.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05015137.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168110.hg.1 | TC0500009671.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168112.hg.1 | TC0500009671.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168114.hg.1 | TC0500009671.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168115.hg.1 | TC0500009671.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500168113.hg.1 | TC0500009671.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000361362.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000520805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | ENST00000523286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009671.hg.1 | NM_030613.3 | Refseq |

ZFP2
Genomic Map