| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038298.hg.1 |
TC03002165.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038298.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038298.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038298.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038297.hg.1 |
TC03002165.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038297.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03038297.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202525.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202525.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202525.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202525.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202525.hg.1 |
TC0300013719.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300202526.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202526.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202526.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202526.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202526.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202526.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202526.hg.1 |
TC0300013719.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300202527.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202527.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202527.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202527.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202527.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202527.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202527.hg.1 |
TC0300013719.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300202529.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202529.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202529.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300202529.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300202529.hg.1 |
TC0300013719.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000399942.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
ENST00000409690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013719.hg.1 |
NM_032898.4 |
Refseq |