Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006451.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006451.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006451.hg.1 |
TC16000464.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600157998.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157998.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157998.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157998.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157998.hg.1 |
TC1600007953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007953.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157999.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157999.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157999.hg.1 |
ENST00000219162.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157999.hg.1 |
NM_032935.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157999.hg.1 |
TC1600007953.hg.1 |
Clariom D TC |