Assay Details
| Gene Symbol: SIGLEC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033329.2 | NP_201586.1 | 1 - 2 | 471 | 89 | |
| NM_053003.3 | NP_443729.1 | 2 - 3 | 861 | 89 | ||
| XM_011527441.2 | XP_011525743.1 | 2 - 3 | 558 | 89 | ||
| XM_011527442.2 | XP_011525744.1 | 2 - 3 | 861 | 89 | ||
| GenBank mRNA | AF282256.1 | - | 2 - 3 | 841 | 89 | |
| AK095919.1 | - | 2 - 3 | 861 | 89 | ||
| AK225931.1 | - | 2 - 3 | 524 | 89 | ||
| AY358140.1 | - | 2 - 3 | 819 | 89 | ||
| BC035809.1 | - | 2 - 3 | 858 | 5755526 | 89 | |
More Information
Set Membership: |
|

SIGLEC12
Genomic Map