Assay Details
| Gene Symbol: TP53RK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033550.3 | NP_291028.3 | 1 - 2 | 508 | 94 | |
| GenBank mRNA | AB017505.2 | - | 1 - 2 | 501 | 94 | |
| AB065434.1 | - | 1 - 2 | 530 | 94 | ||
| AK096502.1 | - | 1 - 2 | 289 | 94 | ||
| AK315182.1 | - | 1 - 2 | 334 | 94 | ||
| BC009727.1 | - | 1 - 2 | 438 | 3899629 | 94 | |
| BC035461.1 | - | 1 - 2 | 282 | 94 | ||
| BC066309.1 | - | 1 - 2 | 304 | 5277911 | 94 | |
More Information
Set Membership: |
|

TP53RK
Genomic Map