| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800006553.hg.1 |
ENST00000520775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054057.hg.1 |
TC01003474.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054057.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054057.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054058.hg.1 |
TC01003474.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054058.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054058.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054059.hg.1 |
TC01003474.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054059.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054056.hg.1 |
TC01003474.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054056.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01054056.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223924.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223924.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223924.hg.1 |
TC0100016290.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100223923.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223923.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223923.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223923.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223923.hg.1 |
TC0100016290.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800006553.hg.1 |
ENST00000520775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223921.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223921.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223921.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223921.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223921.hg.1 |
TC0100016290.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
ENST00000467021.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016290.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223920.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223920.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223920.hg.1 |
ENST00000367874.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100223920.hg.1 |
NM_053053.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100223920.hg.1 |
TC0100016290.hg.1 |
Clariom D TC |