Assay Details
Gene Symbol: DUSP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_111940.1 | 1 - 2 | 190 | 79 | ||
GenBank mRNA | AK022513.1 | - | 1 - 2 | 103 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK022513.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
regulation of adaptive immune response
protein dephosphorylation
response to stress
JNK cascade
dephosphorylation
response to lipopolysaccharide
peptidyl-tyrosine dephosphorylation
negative regulation of JUN kinase activity
negative regulation of protein kinase activity by regulation of protein phosphorylation
oligodendrocyte differentiation
negative regulation of oligodendrocyte differentiation
negative regulation of respiratory burst involved in inflammatory response
regulation of brown fat cell differentiation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
MAP kinase phosphatase activity
inactivation of MAPK activity
regulation of adaptive immune response
protein dephosphorylation
response to stress
JNK cascade
dephosphorylation
response to lipopolysaccharide
peptidyl-tyrosine dephosphorylation
negative regulation of JUN kinase activity
negative regulation of protein kinase activity by regulation of protein phosphorylation
oligodendrocyte differentiation
negative regulation of oligodendrocyte differentiation
negative regulation of respiratory burst involved in inflammatory response
regulation of brown fat cell differentiation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
MAP kinase phosphatase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000494642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000494642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | ENST00000494642.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231677.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231677.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231677.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231677.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231677.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231677.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231677.hg.1 | TC0100017420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000468085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000494642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000494642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | ENST00000494642.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017420.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | ENST00000366899.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | ENST00000477026.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | NM_007207.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | NR_111940.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231674.hg.1 | TC0100017420.hg.1 | Clariom D TC |
