Assay Details
| Gene Symbol: SPATA31D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145197.1 | NP_001138669.1 | 1 - 2 | 277 | 94 | |
| GenBank mRNA | AK125847.1 | - | 1 - 2 | 222 | 94 | |
| AK308327.1 | - | 1 - 2 | 218 | 94 | ||
| Gene Symbol: SPATA31D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207416.2 | NP_997299.2 | 1 - 2 | 277 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000366.hg.1, TC09000367.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09004012.hg.1 | TC09000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004012.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004013.hg.1 | TC09000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004013.hg.1 | ENST00000341875.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004013.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004018.hg.1 | TC09000367.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004018.hg.1 | ENST00000334208.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004018.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004014.hg.1 | TC09000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004014.hg.1 | ENST00000341875.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004014.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004017.hg.1 | TC09000367.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004017.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004019.hg.1 | TC09000367.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004019.hg.1 | ENST00000334208.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004019.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152105.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152105.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152105.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152105.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152105.hg.1 | TC0900007710.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900184187.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184187.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184187.hg.1 | TC0900012145.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012145.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152104.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152104.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152104.hg.1 | TC0900007710.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900184188.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184188.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184188.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184188.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184188.hg.1 | TC0900012145.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900152106.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152106.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152106.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900152106.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900152106.hg.1 | TC0900007710.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900184189.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184189.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184189.hg.1 | ENST00000445385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184189.hg.1 | NM_207416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184189.hg.1 | TC0900012145.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000344803.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | ENST00000603921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012146.hg.1 | NM_001001670.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | ENST00000419782.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007710.hg.1 | NM_001145197.1 | Refseq |

SPATA31D4
Genomic Map