Assay Details
| Gene Symbol: PRDM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522829.1 | XP_011521131.1 | 9 - 10 | 1154 | 90 | |
| GenBank mRNA | AF274348.1 | - | 2 - 3 | 478 | 90 | |
| AL137711.1 | - | 2 - 3 | 303 | 90 | ||
| BC107033.2 | - | 2 - 3 | 376 | 90 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000569206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000569206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | ENST00000569206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000325921.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000449207.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000564210.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000568473.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000569206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000569206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | ENST00000569206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011306.hg.1 | NM_001098173.1 | Refseq |

PRDM7
Genomic Map