| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
ENST00000613087.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
NM_172139.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
NM_172139.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
NM_172139.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
NM_172139.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010646.hg.1 |
NM_172139.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
ENST00000331982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
ENST00000331982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
ENST00000331982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
ENST00000331982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
ENST00000331982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
ENST00000331982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
ENST00000331982.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
NM_172138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
NM_172138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
NM_172138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
NM_172138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
NM_172138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
NM_172138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008050.hg.1 |
NM_172138.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010645.hg.1 |
ENST00000595082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010645.hg.1 |
ENST00000595082.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010645.hg.1 |
ENST00000595082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010645.hg.1 |
ENST00000595082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010645.hg.1 |
ENST00000595082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010645.hg.1 |
ENST00000595082.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900187827.hg.1 |
ENST00000413851.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900187827.hg.1 |
TC1900010646.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900187827.hg.1 |
ENST00000413851.2 |
Ensembl |