| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002407.hg.1 |
ENST00000375500.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002407.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002407.hg.1 |
TC10000203.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014467.hg.1 |
ENST00000375318.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014467.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014467.hg.1 |
TC10001152.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002405.hg.1 |
ENST00000375500.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002405.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002405.hg.1 |
TC10000203.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002406.hg.1 |
ENST00000375500.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002406.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002406.hg.1 |
TC10000203.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014465.hg.1 |
ENST00000375318.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014465.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014465.hg.1 |
TC10001152.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002404.hg.1 |
ENST00000375500.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002404.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002404.hg.1 |
TC10000203.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014466.hg.1 |
ENST00000375318.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014466.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10014466.hg.1 |
TC10001152.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002408.hg.1 |
ENST00000375500.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002408.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10002408.hg.1 |
TC10000203.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000148727.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148727.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148727.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148727.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148727.hg.1 |
TC1000007144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000169550.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000169550.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000169550.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000169550.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000169550.hg.1 |
TC1000010211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000169549.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000169549.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000169549.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000169549.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000169549.hg.1 |
TC1000010211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000148728.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148728.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148728.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148728.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148728.hg.1 |
TC1000007144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000007144.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148725.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148725.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148725.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148725.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148725.hg.1 |
TC1000007144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000148730.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148730.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148730.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148730.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148730.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148730.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148730.hg.1 |
TC1000007144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000148729.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148729.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148729.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148729.hg.1 |
ENST00000375500.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148729.hg.1 |
ENST00000494304.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000148729.hg.1 |
NM_032517.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000148729.hg.1 |
TC1000007144.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000169548.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000169548.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000169548.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000169548.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000169548.hg.1 |
TC1000010211.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
ENST00000375318.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010211.hg.1 |
NM_183058.2 |
Refseq |