Assay Details
| Gene Symbol: AKR1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135241.2 | NP_001128713.1 | 3 - 4 | 543 | 115 | |
| GenBank mRNA | AK296686.1 | - | 3 - 4 | 390 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000986.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
prostaglandin metabolic process
G-protein coupled receptor signaling pathway
digestion
steroid metabolic process
positive regulation of cell proliferation
epithelial cell differentiation
progesterone metabolic process
daunorubicin metabolic process
doxorubicin metabolic process
positive regulation of protein kinase B signaling
oxidation-reduction process
cellular response to jasmonic acid stimulus
cellular response to prostaglandin D stimulus
alditol:NADP+ 1-oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
phenanthrene 9,10-monooxygenase activity
carboxylic acid binding
bile acid binding
ketosteroid monooxygenase activity
trans-1,2-dihydrobenzene-1,2-diol dehydrogenase activity
prostaglandin metabolic process
G-protein coupled receptor signaling pathway
digestion
steroid metabolic process
positive regulation of cell proliferation
epithelial cell differentiation
progesterone metabolic process
daunorubicin metabolic process
doxorubicin metabolic process
positive regulation of protein kinase B signaling
oxidation-reduction process
cellular response to jasmonic acid stimulus
cellular response to prostaglandin D stimulus
alditol:NADP+ 1-oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
phenanthrene 9,10-monooxygenase activity
carboxylic acid binding
bile acid binding
ketosteroid monooxygenase activity
trans-1,2-dihydrobenzene-1,2-diol dehydrogenase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000380753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000421196.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000455190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000460124.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000603073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000603073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000603073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000603073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604184.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604428.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | ENST00000604711.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001135241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_001354.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012516.hg.1 | NM_205845.2 | Refseq |

AKR1C2
Genomic Map