Assay Details
| Gene Symbol: LILRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278404.2 | NP_001265333.2 | 7 | 1396 | 60 | |
| NM_001278405.2 | NP_001265334.2 | 7 | 1329 | 60 | ||
| NM_001278406.2 | NP_001265335.2 | 7 | 1329 | 60 | ||
| NM_005874.4 | NP_005865.3 | 8 | 1547 | 60 | ||
| GenBank mRNA | AF000574.1 | - | 7 | 1281 | 60 | |
| AF011565.1 | - | 7 | 1283 | 60 | ||
| AF011566.1 | - | 7 | 1283 | 60 | ||
| AF025528.1 | - | 8 | 1464 | 60 | ||
| AF283987.1 | - | 7 | 1330 | 60 | ||
| AK297041.1 | - | 7 | 1374 | 60 | ||
| BC036827.1 | - | 8 | 1437 | 5212543 | 60 | |
| EU915606.1 | - | 7 | 1281 | 60 | ||
| LC125264.1 | - | 6 | 1233 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LILRB2
Genomic Map