Assay Details
Gene Symbol: MT1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005952.3 | NP_005943.1 | 1 | 52 | 105 | |
GenBank mRNA | BC018190.1 | - | 1 | 37 | 3859643 | 105 |
BC032131.2 | - | 1 | 41 | 4053295 | 105 | |
BC032338.1 | - | 1 | 39 | 5190391 | 105 | |
CD101947.1 | - | 1 | 56 | 105 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000476.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC018190.1,BC032131.2,BC032338.1,NM_005952.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR16006542.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16006542.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006542.hg.1 | TC16000476.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100013037.hg.1 | ENST00000421314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013037.hg.1 | ENST00000421314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010885.hg.1 | ENST00000396160.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16006543.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16006543.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16006543.hg.1 | TC16000476.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013037.hg.1 | ENST00000421314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013037.hg.1 | ENST00000421314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011399.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600190642.hg.1 | TC1600011399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600190644.hg.1 | TC1600011399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600190645.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190645.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190645.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190645.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190645.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190645.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190645.hg.1 | TC1600011399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000394485.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000562939.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000564974.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | ENST00000568370.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | NM_005952.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600190641.hg.1 | TC1600011399.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900010885.hg.1 | ENST00000396160.4 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 119685 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 173072 | - | - |
