Assay Details
| Gene Symbol: PCDH11X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168361.1 | NP_001161833.1 | 6 - 7 | 3252 | 72 | |
| GenBank mRNA | AY861435.1 | - | 6 - 7 | 3208 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000454.hg.1
Set Membership: |
|

PCDH11X
Genomic Map