Assay Details
Gene Symbol: ZNF492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020855.2 | NP_065906.1 | 1 | 48 | 111 | |
GenBank mRNA | BP871882.1 | - | 1 | 48 | 111 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000400.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_020855.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19005328.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19005328.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19005328.hg.1 | TC19000400.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900009935.hg.1 | ENST00000439134.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009935.hg.1 | ENST00000439134.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000442497.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19005331.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19005331.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19005331.hg.1 | TC19000400.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19005332.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19005332.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19005332.hg.1 | TC19000400.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900007571.hg.1 | ENST00000594711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19005329.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19005329.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19005329.hg.1 | TC19000400.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19005330.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19005330.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19005330.hg.1 | TC19000400.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000397104.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000442497.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | ENST00000596209.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010195.hg.1 | NM_001080409.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155731.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155731.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155731.hg.1 | TC1900007570.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900155731.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155731.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155732.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155732.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155732.hg.1 | TC1900007570.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900155732.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155732.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155728.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155728.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155728.hg.1 | TC1900007570.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900155728.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155728.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155729.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155729.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155729.hg.1 | TC1900007570.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900155729.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155729.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007570.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155730.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155730.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900155730.hg.1 | TC1900007570.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900155730.hg.1 | ENST00000456783.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900155730.hg.1 | NM_020855.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009935.hg.1 | ENST00000439134.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009935.hg.1 | ENST00000439134.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007571.hg.1 | ENST00000594711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000357774.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000593657.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | ENST00000601553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011920.hg.1 | NM_001098626.1 | Refseq |
