Assay Details
| Gene Symbol: MRPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023937.3 | NP_076426.1 | 1 | 246 | 82 | |
| XM_011528189.2 | XP_011526491.1 | 2 | 211 | 82 | ||
| GenBank mRNA | AB049652.1 | - | 1 | 84 | 82 | |
| BC000071.2 | - | 1 | 76 | 3504266 | 82 | |
| BC021801.1 | - | 1 | 245 | 3848952 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000331.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB049652.1,BC000071.2,BC021801.1,NM_023937.3,XM_011528189.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19004337.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004337.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004337.hg.1 | TC19000331.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004341.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004341.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004341.hg.1 | TC19000331.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004340.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004340.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004340.hg.1 | TC19000331.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004339.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004339.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19004339.hg.1 | TC19000331.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153210.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153203.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153203.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153203.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153203.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153203.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153211.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153214.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153214.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153214.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153214.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153214.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153214.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153214.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153215.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153215.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153215.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153215.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153215.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153213.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153207.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000597996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | ENST00000602206.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007319.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153205.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | NM_023937.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | TC1900007319.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000252602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000594999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000595444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | ENST00000600434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900153209.hg.1 | NM_023937.3 | Refseq |

MRPL34
Genomic Map