Assay Details
| Gene Symbol: ATP5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006886.3 | NP_008817.1 | 1 | 53 | 99 | |
| GenBank mRNA | BC105811.1 | - | 1 | 47 | 6739745 | 99 |
| CB106920.1 | - | 1 | 68 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001753.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC105811.1,NM_006886.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20013539.hg.1 | TC20001753.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013539.hg.1 | ENST00000243997.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013539.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013539.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000355937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000371033.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000395663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000463057.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | ENST00000466051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_001256403.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NM_016045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037929.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009648.hg.1 | NR_037930.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167320.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167320.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167320.hg.1 | NM_006886.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167320.hg.1 | TC2000009648.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000167320.hg.1 | ENST00000243997.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167320.hg.1 | ENST00000395659.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167320.hg.1 | NM_006886.3 | Refseq |

ATP5E
Genomic Map