Assay Details
| Gene Symbol: SRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130440.1 | NP_001123912.1 | 1 | 53 | 93 | |
| NM_003133.5 | NP_003124.1 | 1 | 53 | 93 | ||
| GenBank mRNA | DA983529.1 | - | 1 | 53 | 93 | |
| DB194289.1 | - | 1 | 58 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001855.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001130440.1,NM_003133.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01028892.hg.1 | TC01001855.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028892.hg.1 | ENST00000304786.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028892.hg.1 | ENST00000366839.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028892.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028892.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011415.hg.1 | ENST00000445874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011415.hg.1 | ENST00000445874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028894.hg.1 | TC01001855.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028894.hg.1 | ENST00000304786.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028894.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028894.hg.1 | ENST00000366839.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028894.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028894.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028895.hg.1 | TC01001855.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028895.hg.1 | ENST00000366839.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028895.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028902.hg.1 | TC01001855.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028902.hg.1 | ENST00000304786.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028902.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028902.hg.1 | ENST00000366839.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028902.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028902.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028893.hg.1 | TC01001855.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028893.hg.1 | ENST00000304786.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028893.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028893.hg.1 | ENST00000366839.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028893.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028893.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186846.hg.1 | TC0100011767.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186855.hg.1 | TC0100011767.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186856.hg.1 | TC0100011767.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011415.hg.1 | ENST00000445874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011415.hg.1 | ENST00000445874.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186843.hg.1 | TC0100011767.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100186847.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186847.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186847.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186847.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186847.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186847.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186847.hg.1 | TC0100011767.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186845.hg.1 | TC0100011767.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000304786.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366838.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000366839.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000619790.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | ENST00000626563.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_001130440.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_003133.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011767.hg.1 | NM_003133.5 | Refseq |

SRP9
Genomic Map