| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007088.hg.1 |
ENST00000622682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012440.hg.1 |
ENST00000298292.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012440.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012440.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012440.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012440.hg.1 |
TC14001103.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166988.hg.1 |
TC1400009106.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009106.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
ENST00000298292.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
ENST00000406043.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
NM_001083908.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
NM_018139.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400166987.hg.1 |
TC1400009106.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400007088.hg.1 |
ENST00000622682.1 |
Ensembl |