Assay Details
Gene Symbol: MIR101-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029516.1 | 1 | 30 | 72 | ||
GenBank mRNA | AF480539.1 | - | 1 | 30 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 3671 | MIR3671 | -592.0 | Sense | Chr. 1 - 65057755 - 65057842 on NCBI Build GRCh38 | |
Downstream | adenylate kinase 4 | AK4 |
AK 4, AK3, AK3L1, AK3L2 |
89.0kb | Anti-sense | Chr. 1 - 65147342 - 65232145 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014471.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002746.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255348 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283609 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255331 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255338 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283598 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255286 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255336 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255343 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255355 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283590 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283605 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283621 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255333 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283591 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15068075 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283606 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283610 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255334 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283602 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283613 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255354 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283619 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283622 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283608 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255345 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255326 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255323 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255351 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255362 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283593 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255360 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283603 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283621 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283591 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283602 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255345 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283640 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255348 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255354 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283610 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283606 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255351 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15068075 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283590 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255343 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283588 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283603 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255323 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283605 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283608 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283598 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283619 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255338 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255286 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283622 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283593 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283617 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283613 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255362 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283639 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255326 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283595 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283630 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255331 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255360 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255333 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20283609 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255334 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255336 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00255355 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7916778 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7916777 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 239130_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01042984.hg.1 | TC01002746.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01042984.hg.1 | ENST00000362265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01042984.hg.1 | NR_029516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014471.hg.1 | ENST00000362265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014471.hg.1 | NR_029516.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100208222.hg.1 | ENST00000362265.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100208222.hg.1 | NR_029516.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100208222.hg.1 | ENST00000362265.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100208222.hg.1 | NR_029516.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100208222.hg.1 | TC0100014471.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100014471.hg.1 | ENST00000362265.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014471.hg.1 | NR_029516.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3309473 | NR_029516.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3309473 | AF480539.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309473 | NR_029516.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309473 | AF480539.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309473 | NR_029516.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309473 | AF480539.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309473 | NR_029516.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309473 | AF480539.1 | human_transcriptome |
