Assay Details
| Gene Symbol: MIR152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029687.1 | 1 | 46 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CDK5 regulatory subunit associated protein 3 | CDK5RAP3 |
C53, HSF-27, IC53, LZAP, MST016, OK/SW-cl.114, PP1553 |
-55.4kb | Anti-sense | Chr. 17 - 47967904 - 47981786 on NCBI Build GRCh38 |
| Downstream |
nuclear factor, erythroid 2 like 1 |
NFE2L1 |
LCR-F1, NRF1, TCF11 |
11.1kb | Anti-sense | Chr. 17 - 48048324 - 48061545 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1700010974.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001634.hg.1
Set Membership: |
|

MIR152
Genomic Map