Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.1 ST Array |
8087252 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024583.hg.1 |
TC03001396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024583.hg.1 |
ENST00000384873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03024583.hg.1 |
NR_029690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011046.hg.1 |
ENST00000384873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011046.hg.1 |
NR_029690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181411.hg.1 |
ENST00000384873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181411.hg.1 |
NR_029690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181411.hg.1 |
ENST00000384873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300181411.hg.1 |
NR_029690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300181411.hg.1 |
TC0300011046.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011046.hg.1 |
ENST00000384873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011046.hg.1 |
NR_029690.1 |
Refseq |