Assay Details
| Gene Symbol: MIR188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029708.1 | 1 | 35 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 532 | MIR532 |
MIRN532, hsa-mir-532, mir-532 |
-265.0 | Sense | Chr. X - 50003148 - 50003238 on NCBI Build GRCh38 |
| Downstream | microRNA 500a | MIR500A |
MIR500, MIRN500, hsa-mir-500, hsa-mir-500a, mir-500a |
4.8kb | Sense | Chr. X - 50008431 - 50008514 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00007242.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000280.hg.1
Set Membership: |
|

MIR188
Genomic Map