Assay Details
| Gene Symbol: MIR371A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029864.1 | 1 | 33 | 62 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 371b | MIR371B | Anti-sense | Chr. 19 - 53787677 - 53787742 on NCBI Build GRCh38 | ||
| Downstream | microRNA 372 | MIR372 |
MIRN372, hsa-mir-372 |
149.0 | Sense | Chr. 19 - 53787890 - 53787956 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1900008813.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000861.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Chr array_18 19 21 | A_17_P10998273 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640021 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998266 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640018 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159570 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159575 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159572 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998267 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640024 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998276 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640020 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640029 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640026 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998268 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159571 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998264 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159574 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640014 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640012 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159573 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640011 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998277 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998265 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640023 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640017 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998263 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998270 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998271 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640013 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640028 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998272 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998269 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998275 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998262 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998278 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998261 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640025 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998274 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640013 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640018 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998270 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159572 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998273 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640014 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998277 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640020 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159571 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998265 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640023 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159570 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640026 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998264 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998262 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159574 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998267 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640016 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998274 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640028 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640011 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640024 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998272 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998269 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640021 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640029 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998278 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998263 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640018 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998266 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998276 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640012 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998275 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998268 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640017 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998261 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159575 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640013 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998270 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31640025 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17159573 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10998271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640013 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640018 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640023 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998273 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640025 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31640014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10998268 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19011532.hg.1 | ENST00000362161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19011532.hg.1 | NR_029864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19011532.hg.1 | TC19000861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008813.hg.1 | ENST00000362161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008813.hg.1 | NR_029864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900168623.hg.1 | ENST00000362161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900168623.hg.1 | NR_029864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900168623.hg.1 | TC1900008813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900168623.hg.1 | ENST00000362161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900168623.hg.1 | NR_029864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008813.hg.1 | ENST00000362161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008813.hg.1 | NR_029864.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308956 | NR_039909.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3308956 | NR_029864.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308956 | NR_039909.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308956 | NR_029864.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308956 | NR_039909.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308956 | NR_029864.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308956 | NR_039909.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308956 | NR_029864.1 | human_transcriptome |

MIR371A
Genomic Map