Assay Details
| Gene Symbol: MIR382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029874.1 | 1 | 28 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 487a | MIR487A |
MIRN487, MIRN487A, hsa-mir-487, mir-487a |
-1.8kb | Sense | Chr. 14 - 101052446 - 101052525 on NCBI Build GRCh38 |
| Downstream | microRNA 134 | MIR134 |
MIRN134, mir-134 |
306.0 | Sense | Chr. 14 - 101054687 - 101054759 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1400008296.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000749.hg.1
Set Membership: |
|

MIR382
Genomic Map