Assay Details
| Gene Symbol: MIR505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030230.1 | 1 | 42 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATPase phospholipid transporting 11C | ATP11C |
ATPIG, ATPIQ |
Sense | Chr. X - 139726346 - 139933083 on NCBI Build GRCh38 | |
| Downstream | chromosome X open reading frame 66 | CXorf66 | SGPX | 31.5kb | Sense | Chr. X - 139955725 - 139965518 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00010969.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001430.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989559 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989530 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989558 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722650 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722677 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722685 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989590 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722664 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722689 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989530 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989535 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722675 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722672 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989559 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11989592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32722724 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019016.hg.1 | TC0X001430.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019016.hg.1 | ENST00000384901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019016.hg.1 | NR_030230.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010969.hg.1 | ENST00000384901.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010969.hg.1 | NR_030230.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174592.hg.1 | ENST00000384901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174592.hg.1 | NR_030230.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174592.hg.1 | ENST00000384901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174592.hg.1 | NR_030230.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174592.hg.1 | TC0X00010969.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010969.hg.1 | ENST00000384901.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010969.hg.1 | NR_030230.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310381 | NR_030230.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310381 | NR_030230.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310381 | NR_030230.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310381 | NR_030230.1 | human_transcriptome |

MIR505
Genomic Map