Assay Details
| Gene Symbol: MIR506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030233.1 | 1 | 65 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | periphilin-1-like | LOC105373347 | Anti-sense | Chr. X - 147181049 - 147271945 on NCBI Build GRCh38 | ||
| Downstream | microRNA 507 | MIR507 |
MIRN507, hsa-mir-507, mir-507 |
141.0 | Sense | Chr. X - 147230984 - 147231077 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0X00011046.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001463.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745609 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010449 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17408613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010448 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17408611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17408612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745609 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745639 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17408612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010448 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17408611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745622 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745637 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17408613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745636 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745644 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745623 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P12010449 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P32745612 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019123.hg.1 | TC0X001463.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019123.hg.1 | ENST00000384998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X019123.hg.1 | NR_030233.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011046.hg.1 | ENST00000384998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011046.hg.1 | NR_030233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174861.hg.1 | ENST00000384998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174861.hg.1 | NR_030233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174861.hg.1 | ENST00000384998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00174861.hg.1 | NR_030233.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00174861.hg.1 | TC0X00011046.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011046.hg.1 | ENST00000384998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011046.hg.1 | NR_030233.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310246 | NR_030233.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310246 | NR_030233.1 | human_transcriptome |

MIR506
Genomic Map