Assay Details
Gene Symbol: MIR591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030322.1 | 1 | 53 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 25 member 13 | SLC25A13 |
ARALAR2, CITRIN, CTLN2 |
Sense | Chr. 7 - 96120220 - 96322147 on NCBI Build GRCh38 | |
Downstream | chromosome 7 open reading frame 76 | C7orf76 | 261.9kb | Sense | Chr. 7 - 96481626 - 96503523 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700011851.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001620.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882753 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631872 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882766 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631865 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631880 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631860 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882770 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882762 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882744 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631887 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882770 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882785 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882791 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882744 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631836 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631880 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882766 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882753 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882780 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882762 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631865 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882767 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631875 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882745 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631887 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882786 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631872 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882778 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631845 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882747 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882788 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882760 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631860 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882783 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05631878 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882771 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25882758 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07024685.hg.1 | TC07001620.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07024685.hg.1 | ENST00000385290.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07024685.hg.1 | NR_030322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011851.hg.1 | ENST00000385290.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011851.hg.1 | NR_030322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184745.hg.1 | ENST00000385290.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184745.hg.1 | NR_030322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184745.hg.1 | ENST00000385290.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184745.hg.1 | NR_030322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184745.hg.1 | TC0700011851.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700011851.hg.1 | ENST00000385290.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011851.hg.1 | NR_030322.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3309069 | NR_030322.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3309069 | NR_030322.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309069 | NR_030322.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309069 | NR_030322.1 | human_transcriptome |
