Assay Details
| Gene Symbol: MIR3138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036090.1 | 1 | 44 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 2 member 9 | SLC2A9 |
GLUT9, GLUTX, UAQTL2, URATv1 |
-38.4kb | Sense | Chr. 4 - 9771125 - 10040248 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 518B | ZNF518B | 361.2kb | Sense | Chr. 4 - 10439880 - 10457427 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0400010034.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001033.hg.1
Set Membership: |
|

MIR3138
Genomic Map