Assay Details
| Gene Symbol: MIR3139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036091.1 | 1 | 33 | 71 | ||
| Gene Symbol: GAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017007969.1 | XP_016863458.1 | 1 | 681 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 38 | USP38 | HP43.8KD | -119.6kb | Sense | Chr. 4 - 143184917 - 143223874 on NCBI Build GRCh38 |
| Downstream | ribosomal protein SA pseudogene 36 | RPSAP36 | RPSA_14_525 | 81.4kb | Sense | Chr. 4 - 143424974 - 143425984 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0400008880.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017007969.1 transcripts.
Set Membership: |
|

MIR3139
Genomic Map