Assay Details
| Gene Symbol: MIR3164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036122.1 | 1 | 42 | 72 | ||
| Gene Symbol: TPCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544805.1 | XP_011543107.1 | 18 | 2425 | 72 | |
| XM_011544806.1 | XP_011543108.1 | 19 | 2475 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MRGPRF antisense RNA 1 | MRGPRF-AS1 | -64.7kb | Sense | Chr. 11 - 69012354 - 69018447 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC338694 | LOC338694 | 64.0kb | Anti-sense | Chr. 11 - 69147228 - 69171562 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1100008219.hg.1
Set Membership: |
|

MIR3164
Genomic Map