Assay Details
| Gene Symbol: MIR3183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036148.1 | 1 | 48 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translocase of inner mitochondrial membrane 22 homolog (yeast) | TIMM22 |
TEX4, TIM22 |
-20.3kb | Anti-sense | Chr. 17 - 997117 - 1002167 on NCBI Build GRCh38 |
| Downstream | basic helix-loop-helix family member a9 | BHLHA9 | BHLHF42 | 248.0kb | Anti-sense | Chr. 17 - 1270564 - 1271271 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1700009350.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000971.hg.1
Set Membership: |
|

MIR3183
Genomic Map