Assay Details
| Gene Symbol: MIR4301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036183.1 | 1 | 40 | 55 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and kinase domain containing 1 | ANKK1 | PKK2 | -49.6kb | Anti-sense | Chr. 11 - 113386014 - 113400418 on NCBI Build GRCh38 |
| Downstream |
transmembrane protease, serine 5 |
TMPRSS5 | SPINESIN | 237.5kb | Sense | Chr. 11 - 113687546 - 113709811 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1100012351.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002303.hg.1
Set Membership: |
|

MIR4301
Genomic Map