Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11025651.hg.1 |
NR_036186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11025651.hg.1 |
TC11002147.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011783.hg.1 |
ENST00000581016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011783.hg.1 |
NR_036186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011783.hg.1 |
ENST00000581016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011783.hg.1 |
NR_036186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190999.hg.1 |
ENST00000581016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190999.hg.1 |
NR_036186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190999.hg.1 |
ENST00000581016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190999.hg.1 |
NR_036186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190999.hg.1 |
TC1100011783.hg.1 |
Clariom D TC |