Assay Details
| Gene Symbol: MIR4308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036194.1 | 1 | 46 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterile alpha motif domain containing 4A | SAMD4A |
SAMD4, SMAUG, SMAUG1, SMG, SMGA |
-84.8kb | Anti-sense | Chr. 14 - 54567612 - 54793315 on NCBI Build GRCh38 |
| Downstream | WD repeat and HMG-box DNA binding protein 1 | WDHD1 |
AND-1, AND1, CHTF4, CTF4 |
60.7kb | Sense | Chr. 14 - 54938938 - 55027105 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1400009240.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14001153.hg.1
Set Membership: |
|

MIR4308
Genomic Map