Assay Details
| Gene Symbol: MIR4255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036217.1 | 1 | 36 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glutamate ionotropic receptor kainate type subunit 3 | GRIK3 |
EAA5, GLR7, GLUR7, GluK3, GluR7a |
-127.0kb | Anti-sense | Chr. 1 - 36795527 - 37034243 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1137 | LINC01137 | 293.2kb | Anti-sense | Chr. 1 - 37454879 - 37474443 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100007827.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000472.hg.1
Set Membership: |
|

MIR4255
Genomic Map