Assay Details
| Gene Symbol: MIR4325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036219.1 | 1 | 49 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bone morphogenetic protein 7 | BMP7 | OP-1 | -54.9kb | Sense | Chr. 20 - 57168753 - 57266651 on NCBI Build GRCh38 |
| Downstream |
SPO11, initiator of meiotic double stranded breaks |
SPO11 |
CT35, SPATA43, TOPVIA |
8.2kb | Anti-sense | Chr. 20 - 57329757 - 57343993 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC2000009585.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000973.hg.1
Set Membership: |
|

MIR4325
Genomic Map