Assay Details
| Gene Symbol: MIR4261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036222.1 | 1 | 31 | 54 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 2 open reading frame 48 | C2orf48 | Anti-sense | Chr. 2 - 10141382 - 10211730 on NCBI Build GRCh38 | ||
| Downstream | hippocalcin like 1 | HPCAL1 |
BDR1, HLP2, VILIP-3 |
110.2kb | Anti-sense | Chr. 2 - 10302124 - 10427617 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200011712.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001568.hg.1
Set Membership: |
|

MIR4261
Genomic Map