Assay Details
| Gene Symbol: SRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321054.1 | NP_001307983.1 | 1 - 2 | 210 | 74 | |
| GenBank mRNA | BQ051861.1 | - | 1 - 2 | 121 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KIF1 binding protein | KIF1BP |
KBP, KIAA1279, TTC20 |
-71.0kb | Sense | Chr. 10 - 68988721 - 69016983 on NCBI Build GRCh38 |
| Downstream |
VPS26, retromer complex component A |
VPS26A |
HB58, Hbeta58, PEP8A, VPS26 |
19.3kb | Sense | Chr. 10 - 69124153 - 69174416 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SRGN
Genomic Map