| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036902.hg.1 |
TC02002300.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036902.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036902.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036901.hg.1 |
TC02002300.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036901.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036901.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000433431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036903.hg.1 |
TC02002300.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036903.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02036903.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203951.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203951.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203951.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203951.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203951.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203951.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203951.hg.1 |
TC0200014243.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000433431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014243.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203953.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203953.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203953.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203953.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203953.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203953.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203953.hg.1 |
TC0200014243.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200203955.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203955.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203955.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203955.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203955.hg.1 |
ENST00000624615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203955.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203955.hg.1 |
TC0200014243.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200203956.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203956.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203956.hg.1 |
ENST00000412425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200203956.hg.1 |
NR_036537.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200203956.hg.1 |
TC0200014243.hg.1 |
Clariom D TC |