Assay Details
| Gene Symbol: RAB6C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036537.1 | 3 - 4 | 468 | 88 | ||
| GenBank mRNA | AK123452.1 | - | 3 - 4 | 405 | 88 | |
| DQ786271.1 | - | 3 - 4 | 474 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927924 | LOC101927924 | -30.4kb | Anti-sense | Chr. 2 - 129923177 - 129946703 on NCBI Build GRCh38 | |
| Downstream |
RAB6C, member RAS oncogene family |
RAB6C | WTH3 | 571.0 | Anti-sense | Chr. 2 - 129979662 - 129982738 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002300.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01478199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15321989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21594032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01478196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01478194 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 30184 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 228814 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 807522 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 521443 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 740958 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 107142 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 106829 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 312756 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 326202 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 106222 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 366354 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 376240 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 228594 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 91133 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 389746 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 320156 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 119608 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P118650 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P118650 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 32184 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45650 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00468230 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00468220 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P35854362 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 9600 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 3063 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 144827 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173900 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 56046 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00468228 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35854362 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P329112 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P13392555 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P35854362 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000433431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036898.hg.1 | TC02002300.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036898.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036898.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036900.hg.1 | TC02002300.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036900.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036900.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000547785.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000547785.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036899.hg.1 | TC02002300.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036899.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02036899.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203948.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203948.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203948.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203948.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203948.hg.1 | TC0200014243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000433431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | ENST00000624615.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014243.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203944.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203944.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203944.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203944.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203944.hg.1 | TC0200014243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200203943.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203943.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203943.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203943.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203943.hg.1 | TC0200014243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200203946.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203946.hg.1 | ENST00000433431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203946.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203946.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203946.hg.1 | ENST00000433431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203946.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203946.hg.1 | TC0200014243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200203949.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203949.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203949.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203949.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203949.hg.1 | TC0200014243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000547785.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000547785.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000548261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000549579.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006475.hg.1 | ENST00000621292.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203945.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203945.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203945.hg.1 | ENST00000412425.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200203945.hg.1 | NR_036537.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200203945.hg.1 | TC0200014243.hg.1 | Clariom D TC |
| Human Genome U95 Set | 50514_r_at | - | - |
| Human Genome U95 Set | 50514_PM_r_at | - | - |
| Human Genome U95 Set | 50513_f_at | - | - |
| Human Genome U95 Set | 50513_PM_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014328 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014327 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014327 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014328 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3282382 | BC044241.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3215992 | BC044241.1 | human_transcriptome |

RAB6C-AS1
Genomic Map