Assay Details
| Gene Symbol: PSMB8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037174.1 | 1 - 2 | 193 | 63 | ||
| GenBank mRNA | BU539272.1 | - | 1 - 2 | 141 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transporter 2, ATP binding cassette subfamily B member |
TAP2 |
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11 |
-5.3kb | Anti-sense | Chr. 6 - 32821833 - 32838823 on NCBI Build GRCh38 |
| Downstream | proteasome subunit beta 9 | PSMB9 |
LMP2, PSMB6i, RING12, beta1i |
7.7kb | Sense | Chr. 6 - 32854161 - 32859851 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004083.hg.1, TC6_apd_hap1000121.hg.1, TC6_cox_hap2000258.hg.1, TC6_dbb_hap3000225.hg.1, TC6_mann_hap4000189.hg.1, TC6_mcf_hap5000201.hg.1, TC6_qbl_hap6000214.hg.1, TC6_ssto_hap7000192.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000667.hg.1 | TC6_apd_hap1000121.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000667.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001388.hg.1 | TC6_mann_hap4000189.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001388.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004694.hg.1 | TC06004083.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004694.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001779.hg.1 | TC6_cox_hap2000258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001779.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001790.hg.1 | TC6_dbb_hap3000225.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001790.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001742.hg.1 | TC6_qbl_hap6000214.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001742.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001601.hg.1 | TC6_ssto_hap7000192.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001601.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001568.hg.1 | TC6_mcf_hap5000201.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001568.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000412095.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000413039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000429600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000453426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014109.hg.1 | NR_037175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198401.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198401.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198401.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198401.hg.1 | ENST00000415067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198401.hg.1 | ENST00000458296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198401.hg.1 | NR_037174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198401.hg.1 | TC0600014109.hg.1 | Clariom D TC |

PSMB8-AS1
Genomic Map