Assay Details
| Gene Symbol: MIR3617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037411.1 | 1 | 40 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WAP four-disulfide core domain 13 | WFDC13 |
C20orf138, WAP13, dJ601O1.3 |
Anti-sense | Chr. 20 - 45702016 - 45708817 on NCBI Build GRCh38 | |
| Downstream |
serine peptidase inhibitor, Kunitz type 4 |
SPINT4 |
C20orf137, SPINT3, dJ601O1.1 |
17.2kb | Anti-sense | Chr. 20 - 45722349 - 45725696 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC2000009240.hg.1
Set Membership: |
|

MIR3617
Genomic Map