Assay Details
| Gene Symbol: MIR3692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037463.1 | 1 | 32 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 242 | TMEM242 |
BM033, C6orf35 |
-204.9kb | Anti-sense | Chr. 6 - 157289022 - 157324259 on NCBI Build GRCh38 |
| Downstream | sorting nexin 9 | SNX9 |
SDP1, SH3PX1, SH3PXD3A, WISP |
186.8kb | Sense | Chr. 6 - 157716046 - 157945077 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0600009986.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001125.hg.1
Set Membership: |
|

MIR3692
Genomic Map