Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055343.hg.1 |
TC01006384.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055343.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055343.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055343.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244235.hg.1 |
TC0100018539.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000415414.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000440959.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
ENST00000622400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018539.hg.1 |
NR_037642.1 |
Refseq |