Assay Details
| Gene Symbol: CES5AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037839.1 | 4 - 5 | 521 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
BCR, RhoGEF and GTPase activating protein |
BCR |
ALL, BCR1, CML, D22S11, D22S662, PHL |
-48.2kb | Anti-sense | Chr. 22 - 23180365 - 23318037 on NCBI Build GRCh38 |
| Downstream | zinc finger DHHC-type containing 8 pseudogene 1 | ZDHHC8P1 | ZDHHC8P | 20.1kb | Sense | Chr. 22 - 23390605 - 23402612 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000558.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3276303 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22010070.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010070.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010070.hg.1 | TC22000558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010066.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010066.hg.1 | TC22000558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010067.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010067.hg.1 | TC22000558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010068.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010068.hg.1 | TC22000558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158353.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158353.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158353.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158353.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158353.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158353.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158353.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158350.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158350.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158350.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158350.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158350.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158362.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158361.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158351.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158351.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158351.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158351.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158351.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Human Genome U95 Set | 89358_at | - | - |
| Human Genome U95 Set | 89358_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1710629 | NR_037839.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1710629 | NR_037839.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1710629 | NR_037839.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1710629 | NR_037839.1 | human_transcriptome |

CES5AP1
Genomic Map